Product Details

SNP ID
rs149950597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140657681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAAAGGTACCTGGAGGGTTAGAG[A/G]GAGAAGCCTTACCCACAGAGGACGT
Phenotype
MIM: 600549 MIM: 602137
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IK PubMed Links

Gene Details

Gene
IK
Gene Name
IK cytokine, down-regulator of HLA II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006083.3 Intron NP_006074.2
Gene
MIR3655
Gene Name
microRNA 3655
There are no transcripts associated with this gene.

Gene
NDUFA2
Gene Name
NADH:ubiquinone oxidoreductase subunit A2
There are no transcripts associated with this gene.

Gene
WDR55
Gene Name
WD repeat domain 55
There are no transcripts associated with this gene.

View Full Product Details