Product Details

SNP ID
rs150142623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:80559116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAGGCTCTTAGCAAATGTGCAG[C/T]GTCTTTTACATCAATGCTTTGGTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD34B PubMed Links

Gene Details

Gene
ANKRD34B
Gene Name
ankyrin repeat domain 34B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004441.2 1334 Missense Mutation ACT,GCT T302A NP_001004441.2
XM_006714599.3 1334 Missense Mutation ACT,GCT T302A XP_006714662.1
XM_011543366.2 1334 Missense Mutation ACT,GCT T302A XP_011541668.1
XM_011543369.2 1334 Missense Mutation ACT,GCT T302A XP_011541671.1

View Full Product Details