Product Details

SNP ID
rs150581386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:173232659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCGCATTCAAGTCCCCGACGCC[C/G]AAGTTCACGAAGTTGTTGTTGGCGG
Phenotype
MIM: 600584
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NKX2-5 PubMed Links

Gene Details

Gene
NKX2-5
Gene Name
NK2 homeobox 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166175.1 1369 UTR 3 NP_001159647.1
NM_001166176.1 1369 UTR 3 NP_001159648.1
NM_004387.3 1369 Silent Mutation TTC,TTG F295L NP_004378.1
XM_017009071.1 1369 Intron XP_016864560.1

View Full Product Details