Product Details

SNP ID
rs150650090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:80558529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTTGCAATGATTGTCTCCTTAA[A/C]AACATTTTCTTACTTTTGAATTCTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD34B PubMed Links

Gene Details

Gene
ANKRD34B
Gene Name
ankyrin repeat domain 34B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004441.2 1921 Missense Mutation TTG,TTT L497F NP_001004441.2
XM_006714599.3 1921 Missense Mutation TTG,TTT L497F XP_006714662.1
XM_011543366.2 1921 Missense Mutation TTG,TTT L497F XP_011541668.1
XM_011543369.2 1921 Missense Mutation TTG,TTT L497F XP_011541671.1

View Full Product Details