Product Details

SNP ID
rs151263831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150847081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGCAGAGCATTTGAGTATGCT[C/G]TCTCCTCCTCTCCCTCACTTCAGTT
Phenotype
MIM: 608212
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IRGM PubMed Links

Gene Details

Gene
IRGM
Gene Name
immunity related GTPase M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145805.1 559 UTR 5 NP_001139277.1
XM_011537641.2 559 UTR 5 XP_011535943.1

View Full Product Details