Product Details

SNP ID
rs17194467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30163808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTTCTGCCGGCTCTGCCTCCCC[A/G]CGCTCTCCCAGATGGGGGCCCAATC
Phenotype
MIM: 605701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM10 PubMed Links

Gene Details

Gene
TRIM10
Gene Name
tripartite motif containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006778.3 603 Intron NP_006769.2
NM_052828.2 603 Intron NP_439893.2
XM_011514221.1 603 Intron XP_011512523.1
XM_011514222.2 603 Intron XP_011512524.1
XM_011514223.2 603 Intron XP_011512525.1
XM_011514225.1 603 Intron XP_011512527.1
Gene
TRIM15
Gene Name
tripartite motif containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033229.2 603 Missense Mutation ACG,GCG T42A NP_150232.2
XM_011514987.1 603 Intron XP_011513289.1
XM_011514988.2 603 UTR 5 XP_011513290.1

View Full Product Details