Product Details

SNP ID
rs137989361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:22287903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACATAGATTTGACGAAGAGACAA[C/T]GGAGTAGCTAGCTTCGGTGCTCTCA
Phenotype
MIM: 176760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRL PubMed Links

Gene Details

Gene
PRL
Gene Name
prolactin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000948.5 Intron NP_000939.1
NM_001163558.2 Intron NP_001157030.1
XM_011514753.2 Intron XP_011513055.1
XM_011514754.2 Intron XP_011513056.1

View Full Product Details