Product Details

SNP ID
rs139217085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36200638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAAGAAGGAATCCTGCTCCAAG[C/T]ATGCATCTGGTACTTCCTTCCACCT
Phenotype
MIM: 616856
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRPF3 PubMed Links

Gene Details

Gene
BRPF3
Gene Name
bromodomain and PHD finger containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015695.2 546 Missense Mutation CAT,TAT H106Y NP_056510.2
XM_005249010.2 546 Missense Mutation CAT,TAT H106Y XP_005249067.1
XM_005249011.3 546 Missense Mutation CAT,TAT H106Y XP_005249068.1
XM_011514489.1 546 Missense Mutation CAT,TAT H106Y XP_011512791.1
XM_011514490.1 546 Missense Mutation CAT,TAT H106Y XP_011512792.1
XM_011514491.1 546 Missense Mutation CAT,TAT H106Y XP_011512793.1
XM_011514492.1 546 Missense Mutation CAT,TAT H106Y XP_011512794.1
XM_017010742.1 546 Missense Mutation CAT,TAT H106Y XP_016866231.1
XM_017010743.1 546 Intron XP_016866232.1
Gene
LOC102723648
Gene Name
uncharacterized LOC102723648
There are no transcripts associated with this gene.

View Full Product Details