Product Details

SNP ID
rs139333181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:96522411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAACTTCTTGCCCTTTTATCTCT[C/T]GTAACAGACCTAGGGCTAGACCCCT
Phenotype
MIM: 613372
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UFL1 PubMed Links

Gene Details

Gene
UFL1
Gene Name
UFM1 specific ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015323.4 Intron NP_056138.1

View Full Product Details