Product Details

SNP ID
rs139647701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33163729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCACCCCTCCCCGCCTCGGTGG[C/G]GCTCCCAGGTCTGAGAAGGAGGCAT
Phenotype
MIM: 120290
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COL11A2 PubMed Links

Gene Details

Gene
COL11A2
Gene Name
collagen type XI alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163771.1 4679 Intron NP_001157243.1
NM_080679.2 4679 Silent Mutation GCC,GCG A1613A NP_542410.2
NM_080680.2 4679 Silent Mutation GCC,GCG A1720A NP_542411.2
NM_080681.2 4679 Silent Mutation GCC,GCG A1634A NP_542412.2
XM_011514299.2 4679 Silent Mutation GCC,GCG A1482A XP_011512601.1
XM_011514300.2 4679 Silent Mutation GCC,GCG A1422A XP_011512602.1
XM_011514302.2 4679 Silent Mutation GCC,GCG A1349A XP_011512604.1
XM_017010250.1 4679 Silent Mutation GCC,GCG A1720A XP_016865739.1
XM_017010251.1 4679 Silent Mutation GCC,GCG A1326A XP_016865740.1

View Full Product Details