Product Details

SNP ID
rs139719967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:22290234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGGTTTGCTCCTCAATCTCTAC[A/G]GCTTTGGATAGGATAGCCTCCGGGG
Phenotype
MIM: 176760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRL PubMed Links

Gene Details

Gene
PRL
Gene Name
prolactin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000948.5 670 Silent Mutation NP_000939.1
NM_001163558.2 670 Silent Mutation NP_001157030.1
XM_011514753.2 670 Silent Mutation XP_011513055.1
XM_011514754.2 670 Silent Mutation XP_011513056.1

View Full Product Details