Product Details

SNP ID
rs139820365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33417521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGACCTTCTCGTTGGGGCAAGTAA[C/G]AAAGGCTGCAGAGACCGAGCCCGGA
Phenotype
MIM: 616953 MIM: 603763 MIM: 602881 MIM: 603384
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CUTA PubMed Links

Gene Details

Gene
CUTA
Gene Name
cutA divalent cation tolerance homolog (E. coli)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014433.2 768 Missense Mutation CTT,GTT L92V NP_001014433.1
NM_001014837.1 768 Missense Mutation CTT,GTT L50V NP_001014837.1
NM_001014838.1 768 Missense Mutation CTT,GTT L50V NP_001014838.1
NM_001014840.1 768 Missense Mutation CTT,GTT L73V NP_001014840.1
NM_015921.2 768 Missense Mutation CTT,GTT L50V NP_057005.1
Gene
KIFC1
Gene Name
kinesin family member C1
There are no transcripts associated with this gene.

Gene
PHF1
Gene Name
PHD finger protein 1
There are no transcripts associated with this gene.

Gene
SYNGAP1
Gene Name
synaptic Ras GTPase activating protein 1
There are no transcripts associated with this gene.

View Full Product Details