Product Details

SNP ID
rs139841919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:4068698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGATGTATGAAAGAAAAGAGTT[A/G]TTTTTGTTCAATGGGTGAGCAGCAC
Phenotype
MIM: 602338
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM217A PubMed Links

Gene Details

Gene
FAM217A
Gene Name
family with sequence similarity 217 member A
There are no transcripts associated with this gene.

Gene
PRPF4B
Gene Name
pre-mRNA processing factor 4B
There are no transcripts associated with this gene.

View Full Product Details