Product Details

SNP ID
rs139865016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:150064701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGGTTGAGCTAAGCCTGGGGCC[A/G]TGGTGGGTGGTGCTGAAAGGGAAAC
Phenotype
MIM: 605699
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ULBP3 PubMed Links

Gene Details

Gene
ULBP3
Gene Name
UL16 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024518.2 722 Missense Mutation NP_078794.1

View Full Product Details