Product Details

SNP ID
rs140210221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132552842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGCTGGCTGTATTTGGAAATCT[C/G]TTAGTAATGACTTCTGTTCTTCATT
Phenotype
MIM: 606927
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TAAR8 PubMed Links

Gene Details

Gene
TAAR8
Gene Name
trace amine associated receptor 8
There are no transcripts associated with this gene.

View Full Product Details