Product Details

SNP ID
rs140242341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42208345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTTTCCCAGATTGGCTCCTTGA[C/T]GTGTTCTGGTAACTGTTCTAATTGT
Phenotype
MIM: 611976
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPS10 PubMed Links

Gene Details

Gene
MRPS10
Gene Name
mitochondrial ribosomal protein S10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018141.3 597 Missense Mutation ATC,GTC I184V NP_060611.2
XM_005249206.2 597 Missense Mutation ATC,GTC I183V XP_005249263.1
XM_011514724.2 597 Missense Mutation ATC,GTC I194V XP_011513026.1
XM_011514725.2 597 Missense Mutation ATC,GTC I193V XP_011513027.1
XM_017010996.1 597 Missense Mutation ATC,GTC I143V XP_016866485.1

View Full Product Details