Product Details
- SNP ID
-
rs140403175
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:83151986 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCTTGAAGTTAAGCCTTCTCCCA[A/G]AATAATGGTAGATGGAACCAATTTG
- Phenotype
-
MIM: 616823
MIM: 172100
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DOPEY1
PubMed Links
Gene Details
- Gene
- DOPEY1
- Gene Name
- dopey family member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001199942.1 |
6236 |
Missense Mutation |
AAA,AGA |
K1994R |
NP_001186871.1 |
NM_015018.3 |
6236 |
Missense Mutation |
AAA,AGA |
K2003R |
NP_055833.2 |
XM_011535619.2 |
6236 |
Missense Mutation |
AAA,AGA |
K1993R |
XP_011533921.1 |
XM_017010559.1 |
6236 |
Missense Mutation |
AAA,AGA |
K2003R |
XP_016866048.1 |
XM_017010560.1 |
6236 |
Missense Mutation |
AAA,AGA |
K2003R |
XP_016866049.1 |
XM_017010561.1 |
6236 |
Missense Mutation |
AAA,AGA |
K1994R |
XP_016866050.1 |
XM_017010562.1 |
6236 |
Missense Mutation |
AAA,AGA |
K1994R |
XP_016866051.1 |
XM_017010563.1 |
6236 |
Missense Mutation |
AAA,AGA |
K2003R |
XP_016866052.1 |
XM_017010564.1 |
6236 |
Missense Mutation |
AAA,AGA |
K2003R |
XP_016866053.1 |
XM_017010565.1 |
6236 |
Missense Mutation |
AAA,AGA |
K1994R |
XP_016866054.1 |
XM_017010566.1 |
6236 |
Missense Mutation |
AAA,AGA |
K1994R |
XP_016866055.1 |
XM_017010567.1 |
6236 |
Missense Mutation |
AAA,AGA |
K1994R |
XP_016866056.1 |
XM_017010568.1 |
6236 |
Missense Mutation |
AAA,AGA |
K1993R |
XP_016866057.1 |
XM_017010569.1 |
6236 |
Missense Mutation |
AAA,AGA |
K1993R |
XP_016866058.1 |
XM_017010570.1 |
6236 |
Missense Mutation |
AAA,AGA |
K1970R |
XP_016866059.1 |
XM_017010571.1 |
6236 |
Missense Mutation |
AAA,AGA |
K1968R |
XP_016866060.1 |
XM_017010572.1 |
6236 |
Missense Mutation |
AAA,AGA |
K2003R |
XP_016866061.1 |
XM_017010573.1 |
6236 |
Missense Mutation |
AAA,AGA |
K2003R |
XP_016866062.1 |
XM_017010574.1 |
6236 |
Missense Mutation |
AAA,AGA |
K2003R |
XP_016866063.1 |
- Gene
- PGM3
- Gene Name
- phosphoglucomutase 3
View Full Product Details