Product Details

SNP ID
rs140510882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116877401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGGGCTTGCTAGTCTATCCGGA[A/T]GAAACAGTGTACCTGGCGGCCGAAG
Phenotype
MIM: 612659
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RFX6 PubMed Links

Gene Details

Gene
RFX6
Gene Name
regulatory factor X6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173560.3 189 Missense Mutation GAA,GAT E42D NP_775831.2
XM_011535589.1 189 Missense Mutation GAA,GAT E42D XP_011533891.1
XM_017010477.1 189 Intron XP_016865966.1

View Full Product Details