Product Details

SNP ID
rs140805833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30619733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATACCAGGAGCGATATGGTTCTC[A/G]CCCCGTCTGGGCTGACTACCGCCGC
Phenotype
MIM: 615556 MIM: 611982 MIM: 603771
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATAT1 PubMed Links

Gene Details

Gene
ATAT1
Gene Name
alpha tubulin acetyltransferase 1
There are no transcripts associated with this gene.

Gene
MRPS18B
Gene Name
mitochondrial ribosomal protein S18B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014046.3 369 Missense Mutation CAC,CGC H71R NP_054765.1
Gene
PPP1R10
Gene Name
protein phosphatase 1 regulatory subunit 10
There are no transcripts associated with this gene.

View Full Product Details