Product Details

SNP ID
rs141012191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49470210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGCAAATGCTTCCAAGAAAAAC[A/G]CTCAACAGTTAAAAAGAAATCCAAA
Phenotype
MIM: 611506 MIM: 609058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CENPQ PubMed Links

Gene Details

Gene
CENPQ
Gene Name
centromere protein Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018132.3 184 Missense Mutation ACT,GCT T12A NP_060602.2
XM_005249205.1 184 Missense Mutation ACT,GCT T12A XP_005249262.1
XM_011514723.2 184 Intron XP_011513025.2
Gene
MUT
Gene Name
methylmalonyl-CoA mutase
There are no transcripts associated with this gene.

View Full Product Details