Product Details

SNP ID
rs141251993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3225759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCAGGACCGAGTCGACCAGCTC[A/G]GCTCCCTCTGTGTAGTGGCCCTTGG
Phenotype
MIM: 612850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TUBB2B PubMed Links

Gene Details

Gene
TUBB2B
Gene Name
tubulin beta 2B class IIb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178012.4 521 Silent Mutation GCC,GCT A110A NP_821080.1

View Full Product Details