Product Details

SNP ID
rs141590731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52900058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTCATTCAAATCTGCCATACCT[C/T]CTGTATACATATCAATTCTGAAAGA
Phenotype
MIM: 605449
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTA3 PubMed Links

Gene Details

Gene
GSTA3
Gene Name
glutathione S-transferase alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000847.4 195 Missense Mutation GAA,GGA E97G NP_000838.3
XM_006715072.3 195 Missense Mutation GAA,GGA E47G XP_006715135.1
XM_011514533.2 195 Missense Mutation GAA,GGA E4G XP_011512835.1
XM_017010794.1 195 Missense Mutation GAA,GGA E47G XP_016866283.1

View Full Product Details