Product Details

SNP ID
rs141609772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:53002274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCACATCAATACCCCTACAACG[C/T]ACCTTCGTCACTGAATTTCTGTCTC
Phenotype
MIM: 605450 MIM: 612325 MIM: 606515
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTA4 PubMed Links

Gene Details

Gene
GSTA4
Gene Name
glutathione S-transferase alpha 4
There are no transcripts associated with this gene.

Gene
ICK
Gene Name
intestinal cell (MAK-like) kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014920.3 5954 UTR 3 NP_055735.1
NM_016513.4 5954 UTR 3 NP_057597.2
XM_011514419.1 5954 UTR 3 XP_011512721.1
XM_011514420.2 5954 UTR 3 XP_011512722.1
XM_011514421.1 5954 UTR 3 XP_011512723.1
XM_017010485.1 5954 UTR 3 XP_016865974.1
XM_017010486.1 5954 UTR 3 XP_016865975.1
XM_017010487.1 5954 UTR 3 XP_016865976.1
XM_017010488.1 5954 UTR 3 XP_016865977.1
XM_017010489.1 5954 UTR 3 XP_016865978.1
XM_017010490.1 5954 UTR 3 XP_016865979.1
XM_017010491.1 5954 UTR 3 XP_016865980.1
XM_017010492.1 5954 UTR 3 XP_016865981.1
Gene
RN7SK
Gene Name
RNA, 7SK small nuclear
There are no transcripts associated with this gene.

View Full Product Details