Product Details

SNP ID
rs141627165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:143941876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCTGCTTTGAGGGGCAGGCTTG[A/C]AAGTGGGGAGTATGAGGTAGAAGTG
Phenotype
MIM: 603044
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLAGL1 PubMed Links

Gene Details

Gene
PLAGL1
Gene Name
PLAG1 like zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080951.2 1623 Missense Mutation NP_001074420.1
NM_001080952.2 1623 Missense Mutation NP_001074421.1
NM_001080953.2 1623 Missense Mutation NP_001074422.1
NM_001080954.2 1623 Missense Mutation NP_001074423.1
NM_001080955.2 1623 Missense Mutation NP_001074424.1
NM_001080956.2 1623 Missense Mutation NP_001074425.1
NM_001289037.1 1623 Missense Mutation NP_001275966.1
NM_001289038.1 1623 Missense Mutation NP_001275967.1
NM_001289039.1 1623 Missense Mutation NP_001275968.1
NM_001289040.1 1623 Missense Mutation NP_001275969.1
NM_001289041.1 1623 Missense Mutation NP_001275970.1
NM_001289042.1 1623 Missense Mutation NP_001275971.1
NM_001289043.1 1623 Missense Mutation NP_001275972.1
NM_001289044.1 1623 Missense Mutation NP_001275973.1
NM_001289045.1 1623 Missense Mutation NP_001275974.1
NM_001289046.1 1623 Missense Mutation NP_001275975.1
NM_001289047.1 1623 Missense Mutation NP_001275976.1
NM_001289048.1 1623 Missense Mutation NP_001275977.1
NM_001289049.1 1623 Missense Mutation NP_001275978.1
NM_001317156.1 1623 Missense Mutation NP_001304085.1
NM_001317157.1 1623 Missense Mutation NP_001304086.1
NM_001317158.1 1623 Missense Mutation NP_001304087.1
NM_001317159.1 1623 Missense Mutation NP_001304088.1
NM_001317160.1 1623 Missense Mutation NP_001304089.1
NM_001317161.1 1623 Missense Mutation NP_001304090.1
NM_001317162.1 1623 Missense Mutation NP_001304091.1
NM_006718.4 1623 Missense Mutation NP_006709.2
Gene
ZC2HC1B
Gene Name
zinc finger C2HC-type containing 1B
There are no transcripts associated with this gene.

View Full Product Details