Product Details

SNP ID
rs142135197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43054364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGGCCAGCCCAGCCCCCCTTGCG[A/G]TGCCACCGCCCACTGTTAGGCCTCT
Phenotype
MIM: 609577 MIM: 611822
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUL7 PubMed Links

Gene Details

Gene
CUL7
Gene Name
cullin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168370.1 813 Intron NP_001161842.1
NM_014780.4 813 Intron NP_055595.2
XM_005249503.2 813 Intron XP_005249560.1
XM_006715285.1 813 Intron XP_006715348.1
XM_011515019.2 813 Intron XP_011513321.1
XM_011515020.2 813 Intron XP_011513322.1
XM_011515021.1 813 Intron XP_011513323.1
XM_017011533.1 813 Intron XP_016867022.1
XM_017011534.1 813 Intron XP_016867023.1
XM_017011535.1 813 Intron XP_016867024.1
XM_017011536.1 813 Intron XP_016867025.1
XM_017011537.1 813 Intron XP_016867026.1
XM_017011538.1 813 Intron XP_016867027.1
XM_017011539.1 813 Intron XP_016867028.1
XM_017011540.1 813 Intron XP_016867029.1
Gene
KLC4
Gene Name
kinesin light chain 4
There are no transcripts associated with this gene.

Gene
MRPL2
Gene Name
mitochondrial ribosomal protein L2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300848.1 813 UTR 3 NP_001287777.1
NM_015950.4 813 Silent Mutation CAC,CAT H276H NP_057034.2
XM_005249161.4 813 Silent Mutation CAC,CAT H244H XP_005249218.1

View Full Product Details