Product Details

SNP ID
rs142454368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43253667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCAGATCTTGGAGTCCATCGAG[G/T]CCATCCACTCTGTGGGCTTCCTGCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TTBK1 PubMed Links

Gene Details

Gene
TTBK1
Gene Name
tau tubulin kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011514947.2 499 Missense Mutation GCC,TCC A144S XP_011513249.1
XM_011514948.2 499 Intron XP_011513250.1
XM_017011364.1 499 Missense Mutation GCC,TCC A144S XP_016866853.1
XM_017011365.1 499 Missense Mutation GCC,TCC A144S XP_016866854.1
XM_017011366.1 499 Missense Mutation GCC,TCC A144S XP_016866855.1
XM_017011367.1 499 Missense Mutation GCC,TCC A144S XP_016866856.1
XM_017011368.1 499 Intron XP_016866857.1

View Full Product Details