Product Details

SNP ID
rs142618598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32170959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAAGCAGATCGAGAGAGCTCTG[A/G]TGGTTGGAGACAACAACATAGGGCT
Phenotype
MIM: 603099 MIM: 609897 MIM: 603298 MIM: 602677
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGPAT1 PubMed Links

Gene Details

Gene
AGPAT1
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006411.3 950 Silent Mutation CAC,CAT H104H NP_006402.1
NM_032741.4 950 Silent Mutation CAC,CAT H104H NP_116130.2
XM_005248805.3 950 Silent Mutation CAC,CAT H108H XP_005248862.1
XM_005248806.2 950 Silent Mutation CAC,CAT H104H XP_005248863.1
XM_011514234.1 950 Silent Mutation CAC,CAT H104H XP_011512536.1
Gene
EGFL8
Gene Name
EGF like domain multiple 8
There are no transcripts associated with this gene.

Gene
MIR6721
Gene Name
microRNA 6721
There are no transcripts associated with this gene.

Gene
MIR6833
Gene Name
microRNA 6833
There are no transcripts associated with this gene.

Gene
PPT2
Gene Name
palmitoyl-protein thioesterase 2
There are no transcripts associated with this gene.

Gene
PPT2-EGFL8
Gene Name
PPT2-EGFL8 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
RNF5
Gene Name
ring finger protein 5
There are no transcripts associated with this gene.

View Full Product Details