Product Details

SNP ID
rs142673484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:125986591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTTAACAGGTATCTGCTCCTCA[A/T]GGCGCAGGAGCATCTGGAGTTCCGC
Phenotype
MIM: 609998
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HINT3 PubMed Links

Gene Details

Gene
HINT3
Gene Name
histidine triad nucleotide binding protein 3
There are no transcripts associated with this gene.

Gene
TRMT11
Gene Name
tRNA methyltransferase 11 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031712.2 63 Intron NP_001026882.2
XM_017011168.1 63 UTR 5 XP_016866657.1
XM_017011169.1 63 UTR 5 XP_016866658.1

View Full Product Details