Product Details

SNP ID
rs142981671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73428741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCTCTTCTTTGTTTTCACAGCA[C/T]GTTTTAGATTTTCCATGATTGTTCA
Phenotype
MIM: 613973
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MB21D1 PubMed Links

Gene Details

Gene
MB21D1
Gene Name
Mab-21 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138441.2 1334 Silent Mutation ACA,ACG T395T NP_612450.2
XM_017010232.1 1334 Silent Mutation ACA,ACG T395T XP_016865721.1

View Full Product Details