Product Details

SNP ID
rs143195021
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43178395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGTGATGTATGCCCCCACCTCG[A/G]GCCTGGGTGATGGCAGCCTCACCGT
Phenotype
MIM: 607489 MIM: 600589
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CUL9 PubMed Links

Gene Details

Gene
CUL9
Gene Name
cullin 9
There are no transcripts associated with this gene.

Gene
SRF
Gene Name
serum response factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292001.1 1626 Missense Mutation AGC,GGC S218G NP_001278930.1
NM_003131.3 1626 Missense Mutation AGC,GGC S422G NP_003122.1

View Full Product Details