Product Details

SNP ID
rs144837997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26544762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAGACAACTTGGTTTTCCTCAG[A/T]TTGATAGATATGAAATGTTATATCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HMGN4 PubMed Links

Gene Details

Gene
HMGN4
Gene Name
high mobility group nucleosomal binding domain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006353.2 Intron NP_006344.1
XM_017010171.1 Intron XP_016865660.1
XM_017010172.1 Intron XP_016865661.1
XM_017010173.1 Intron XP_016865662.1

View Full Product Details