Product Details

SNP ID
rs144935133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109444928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCTCGGCCTCCTCAGCCACCAG[G/T]CTGTTTTTCTTGTCAACGAGCTGTA
Phenotype
MIM: 607129 MIM: 603498
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MICAL1 PubMed Links

Gene Details

Gene
MICAL1
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159291.1 3297 Missense Mutation AGA,AGC R897S NP_001152763.1
NM_001286613.1 3297 Missense Mutation AGA,AGC R1002S NP_001273542.1
NM_022765.3 3297 Missense Mutation AGA,AGC R983S NP_073602.3
Gene
PPIL6
Gene Name
peptidylprolyl isomerase like 6
There are no transcripts associated with this gene.

Gene
SMPD2
Gene Name
sphingomyelin phosphodiesterase 2
There are no transcripts associated with this gene.

View Full Product Details