Product Details

SNP ID
rs145017609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:83554099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCAAATACACTACAATTTCAAGA[G/T]TAGGGATAACAGCTAAGGTAGTACA
Phenotype
MIM: 607923
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SNAP91 PubMed Links

Gene Details

Gene
SNAP91
Gene Name
synaptosome associated protein 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242792.1 2958 UTR 3 NP_001229721.1
NM_001242793.1 2958 UTR 3 NP_001229722.1
NM_001242794.1 2958 UTR 3 NP_001229723.1
NM_001256717.1 2958 UTR 3 NP_001243646.1
NM_001256718.1 2958 UTR 3 NP_001243647.1
NM_014841.2 2958 UTR 3 NP_055656.1
XM_005248770.4 2958 UTR 3 XP_005248827.1
XM_006715615.1 2958 UTR 3 XP_006715678.1
XM_011536265.1 2958 UTR 3 XP_011534567.1
XM_011536266.1 2958 UTR 3 XP_011534568.1
XM_011536269.1 2958 UTR 3 XP_011534571.1
XM_011536271.1 2958 UTR 3 XP_011534573.1
XM_011536273.2 2958 UTR 3 XP_011534575.1
XM_011536275.2 2958 UTR 3 XP_011534577.1
XM_011536276.2 2958 UTR 3 XP_011534578.1
XM_011536277.1 2958 UTR 3 XP_011534579.1
XM_017011554.1 2958 UTR 3 XP_016867043.1
XM_017011555.1 2958 UTR 3 XP_016867044.1
XM_017011556.1 2958 UTR 3 XP_016867045.1
XM_017011557.1 2958 UTR 3 XP_016867046.1
XM_017011558.1 2958 UTR 3 XP_016867047.1
XM_017011559.1 2958 UTR 3 XP_016867048.1
XM_017011560.1 2958 UTR 3 XP_016867049.1
XM_017011561.1 2958 UTR 3 XP_016867050.1
XM_017011562.1 2958 UTR 3 XP_016867051.1
XM_017011563.1 2958 UTR 3 XP_016867052.1
XM_017011564.1 2958 UTR 3 XP_016867053.1
XM_017011565.1 2958 UTR 3 XP_016867054.1
XM_017011566.1 2958 UTR 3 XP_016867055.1
XM_017011567.1 2958 UTR 3 XP_016867056.1
XM_017011568.1 2958 UTR 3 XP_016867057.1
XM_017011569.1 2958 UTR 3 XP_016867058.1
XM_017011570.1 2958 UTR 3 XP_016867059.1
XM_017011571.1 2958 UTR 3 XP_016867060.1
XM_017011572.1 2958 UTR 3 XP_016867061.1
XM_017011573.1 2958 UTR 3 XP_016867062.1
XM_017011574.1 2958 UTR 3 XP_016867063.1
XM_017011575.1 2958 UTR 3 XP_016867064.1
XM_017011576.1 2958 UTR 3 XP_016867065.1
XM_017011577.1 2958 UTR 3 XP_016867066.1
XM_017011578.1 2958 UTR 3 XP_016867067.1
XM_017011579.1 2958 UTR 3 XP_016867068.1
XM_017011580.1 2958 UTR 3 XP_016867069.1
XM_017011581.1 2958 UTR 3 XP_016867070.1
XM_017011582.1 2958 UTR 3 XP_016867071.1
XM_017011583.1 2958 UTR 3 XP_016867072.1
XM_017011584.1 2958 UTR 3 XP_016867073.1
XM_017011585.1 2958 UTR 3 XP_016867074.1
XM_017011586.1 2958 UTR 3 XP_016867075.1
XM_017011587.1 2958 UTR 3 XP_016867076.1
XM_017011588.1 2958 UTR 3 XP_016867077.1
XM_017011589.1 2958 UTR 3 XP_016867078.1
XM_017011590.1 2958 UTR 3 XP_016867079.1

View Full Product Details