Product Details

SNP ID
rs145474393
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:88048080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCGAAAACAACGACAGCGAGACC[A/G]CGGAGAACTACGCTCCGCCTGAAAC
Phenotype
MIM: 612739
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SPACA1 PubMed Links

Gene Details

Gene
SPACA1
Gene Name
sperm acrosome associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030960.2 292 Missense Mutation ACG,GCG T59A NP_112222.1
XM_011536160.2 292 Intron XP_011534462.1
XM_017011335.1 292 Intron XP_016866824.1

View Full Product Details