Product Details

SNP ID
rs145580280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:106364584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAAAGATAGAGTTGCTGCCCTC[C/T]TGAGTCAGCCCCATCCCCTGTCTAG
Phenotype
MIM: 601797
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIM1 PubMed Links

Gene Details

Gene
AIM1
Gene Name
absent in melanoma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001624.3 Intron NP_001615.2
XM_005266839.3 Intron XP_005266896.1
XM_017010333.1 Intron XP_016865822.1

View Full Product Details