Product Details

SNP ID
rs146960695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42923128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTGCCTGGTCCTTGATGTTGCA[C/T]CCCCTGGCCTTGACAGCCCCATCTG
Phenotype
MIM: 610774 MIM: 606817
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNPY3 PubMed Links

Gene Details

Gene
CNPY3
Gene Name
canopy FGF signaling regulator 3
There are no transcripts associated with this gene.

Gene
CNPY3-GNMT
Gene Name
CNPY3-GNMT readthrough
There are no transcripts associated with this gene.

Gene
PTCRA
Gene Name
pre T-cell antigen receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243168.1 241 Missense Mutation CCC,TCC P54S NP_001230097.1
NM_001243169.1 241 Intron NP_001230098.1
NM_001243170.1 241 Intron NP_001230099.1
NM_138296.2 241 Missense Mutation CCC,TCC P54S NP_612153.2

View Full Product Details