Product Details

SNP ID
rs147519555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26545055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACACGAATTATTAACAATATAGT[C/G]AATTTGATTTTTAACGTTTACATTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HMGN4 PubMed Links

Gene Details

Gene
HMGN4
Gene Name
high mobility group nucleosomal binding domain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006353.2 Intron NP_006344.1
XM_017010171.1 Intron XP_016865660.1
XM_017010172.1 Intron XP_016865661.1
XM_017010173.1 Intron XP_016865662.1

View Full Product Details