Product Details

SNP ID
rs147611390
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:54308182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAGCCATAATCTGGGCCATGTA[G/T]ATTCTAAGGAGGCTTTCGATTTAAG
Phenotype
MIM: 606749
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
TINAG PubMed Links

Gene Details

Gene
TINAG
Gene Name
tubulointerstitial nephritis antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014464.3 Intron NP_055279.3
XM_006715062.2 Intron XP_006715125.1
XM_011514497.2 Intron XP_011512799.1
XM_017010745.1 Intron XP_016866234.1
XM_017010746.1 Intron XP_016866235.1
XM_017010747.1 Intron XP_016866236.1
XM_017010748.1 Intron XP_016866237.1

View Full Product Details