Product Details

SNP ID
rs148730821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:108043703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAACTATGTGATTTCATTTCAAT[C/T]GTATCACTTTCATCTGATTGAAATA
Phenotype
MIM: 607649
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OSTM1 PubMed Links

Gene Details

Gene
OSTM1
Gene Name
osteopetrosis associated transmembrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014028.3 2173 UTR 3 NP_054747.2

View Full Product Details