Product Details
- SNP ID
-
rs149044401
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:150876156 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAAAAAAACCCTGCCTTCAAGCC[A/G]GTTCTTGCAATTATCCAGGTAAGCC
- Phenotype
-
MIM: 611427
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MTHFD1L
PubMed Links
Gene Details
- Gene
- MTHFD1L
- Gene Name
- methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001242767.1 |
176 |
Silent Mutation |
CCA,CCG |
P98P |
NP_001229696.1 |
NM_001242768.1 |
176 |
Silent Mutation |
CCA,CCG |
P32P |
NP_001229697.1 |
NM_001242769.1 |
176 |
Silent Mutation |
CCA,CCG |
P98P |
NP_001229698.1 |
NM_015440.4 |
176 |
Silent Mutation |
CCA,CCG |
P98P |
NP_056255.2 |
XM_005266907.4 |
176 |
Silent Mutation |
CCA,CCG |
P98P |
XP_005266964.1 |
XM_005266910.4 |
176 |
Silent Mutation |
CCA,CCG |
P98P |
XP_005266967.1 |
XM_005266911.4 |
176 |
Silent Mutation |
CCA,CCG |
P98P |
XP_005266968.1 |
XM_011535729.2 |
176 |
Silent Mutation |
CCA,CCG |
P98P |
XP_011534031.1 |
XM_011535730.2 |
176 |
Silent Mutation |
CCA,CCG |
P32P |
XP_011534032.1 |
XM_011535731.2 |
176 |
Silent Mutation |
CCA,CCG |
P32P |
XP_011534033.1 |
XM_011535732.2 |
176 |
UTR 5 |
|
|
XP_011534034.1 |
XM_011535733.2 |
176 |
UTR 5 |
|
|
XP_011534035.1 |
XM_011535734.2 |
176 |
UTR 5 |
|
|
XP_011534036.1 |
XM_011535737.2 |
176 |
Silent Mutation |
CCA,CCG |
P98P |
XP_011534039.1 |
XM_011535738.2 |
176 |
Silent Mutation |
CCA,CCG |
P98P |
XP_011534040.1 |
XM_017010702.1 |
176 |
Silent Mutation |
CCA,CCG |
P98P |
XP_016866191.1 |
XM_017010703.1 |
176 |
Silent Mutation |
CCA,CCG |
P98P |
XP_016866192.1 |
XM_017010704.1 |
176 |
Silent Mutation |
CCA,CCG |
P32P |
XP_016866193.1 |
XM_017010705.1 |
176 |
UTR 5 |
|
|
XP_016866194.1 |
XM_017010706.1 |
176 |
UTR 5 |
|
|
XP_016866195.1 |
XM_017010707.1 |
176 |
UTR 5 |
|
|
XP_016866196.1 |
XM_017010708.1 |
176 |
Silent Mutation |
CCA,CCG |
P98P |
XP_016866197.1 |
View Full Product Details