Product Details
- SNP ID
-
rs149532033
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:25653387 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTATTTTCTCACATTTAGAAAAA[G/T]GTTACATAGAAGAGAAGGAACTCGA
- Phenotype
-
MIM: 609202
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SCGN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs60502981] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SCGN
- Gene Name
- secretagogin, EF-hand calcium binding protein
View Full Product Details