Product Details

SNP ID
rs149913884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:99920991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATATTGTTGATTTCCTTCTCAGT[C/T]GCTCTTCTTTGGATTTGAGGCAGAC
Phenotype
MIM: 606111
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCHR2 PubMed Links

Gene Details

Gene
MCHR2
Gene Name
melanin concentrating hormone receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040179.1 1287 Silent Mutation GCA,GCG A324A NP_001035269.1
NM_032503.2 1287 Silent Mutation GCA,GCG A324A NP_115892.2

View Full Product Details