Product Details

SNP ID
rs150711198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:57100689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATGTTCCTCTTCCCATTGGAGA[A/T]AGCAGCTCCTTCTCTGGGAGTTGTT
Phenotype
MIM: 615708
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF451 PubMed Links

Gene Details

Gene
ZNF451
Gene Name
zinc finger protein 451
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031623.2 274 Intron NP_001026794.1
NM_001257273.1 274 Missense Mutation GAA,GAT E91D NP_001244202.1
NM_015555.2 274 Intron NP_056370.2
XM_005248994.1 274 Intron XP_005249051.1
XM_011514460.1 274 Intron XP_011512762.1
XM_011514461.1 274 Intron XP_011512763.1
XM_011514462.2 274 Intron XP_011512764.1
XM_011514463.2 274 Missense Mutation GAA,GAT E63D XP_011512765.1

View Full Product Details