Product Details

SNP ID
rs150721968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29355520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCTCTGAGGACTGGATCCTCA[A/G]GATGGTGGAGATTATGCAAATGTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5V1 PubMed Links

Gene Details

Gene
OR5V1
Gene Name
olfactory receptor family 5 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030876.5 933 Silent Mutation NP_110503.3
XM_011514929.2 933 Missense Mutation XP_011513231.1
XM_011514930.2 933 Missense Mutation XP_011513232.1

View Full Product Details