Product Details

SNP ID
rs150899490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132570627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGAGCTGCTGGTATTTTGGGAG[A/G]AGTTTTTGTACTTTCCACACCTGCT
Phenotype
MIM: 608923
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAAR6 PubMed Links

Gene Details

Gene
TAAR6
Gene Name
trace amine associated receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175067.1 306 Silent Mutation AGA,AGG R102R NP_778237.1

View Full Product Details