Product Details

SNP ID
rs151169403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46704564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAAAAACCTATTTTAATCCTAA[A/T]TGTATTTCTCTATTCCTGAAGAGTT
Phenotype
MIM: 601690 MIM: 611200
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G7 PubMed Links

Gene Details

Gene
PLA2G7
Gene Name
phospholipase A2 group VII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168357.1 1500 Missense Mutation AAT,ATT N441I NP_001161829.1
NM_005084.3 1500 Missense Mutation AAT,ATT N441I NP_005075.3
XM_005249408.4 1500 Missense Mutation AAT,ATT N441I XP_005249465.1
Gene
TDRD6
Gene Name
tudor domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010870.2 1500 Intron NP_001010870.1
NM_001168359.1 1500 Intron NP_001161831.1

View Full Product Details