Product Details

SNP ID
rs151276683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35455955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGTGGGGACCTCCATGGAGGGA[A/G]CTTCTCCACTGTCTGAAAGATGCCA
Phenotype
MIM: 613976
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FANCE PubMed Links

Gene Details

Gene
FANCE
Gene Name
Fanconi anemia complementation group E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021922.2 664 Missense Mutation ACT,GCT T153A NP_068741.1
XM_005248885.2 664 Missense Mutation ACT,GCT T153A XP_005248942.1
XM_005248886.2 664 Missense Mutation ACT,GCT T153A XP_005248943.1
XM_005248887.2 664 Missense Mutation ACT,GCT T153A XP_005248944.1
XM_005248888.3 664 Missense Mutation ACT,GCT T153A XP_005248945.1
XM_011514343.2 664 Missense Mutation ACT,GCT T55A XP_011512645.1

View Full Product Details