Product Details
- SNP ID
-
rs138433522
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:83365839 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTGTAAGGCCCCTGTGATCTGTG[G/T]TTGTGGTTCTATTTCTCCAAATTGA
- Phenotype
-
MIM: 608166
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SEMA3E
PubMed Links
Gene Details
- Gene
- SEMA3E
- Gene Name
- semaphorin 3E
View Full Product Details