Product Details
- SNP ID
-
rs138633855
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:36855587 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATAGACGAAGTCATAGTTGCTGGG[C/T]TCCTTGGGAATCGGCGGAGGTGCGT
- Phenotype
-
MIM: 606420
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ELMO1
PubMed Links
Gene Details
- Gene
- ELMO1
- Gene Name
- engulfment and cell motility 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001039459.2 |
3922 |
Silent Mutation |
GAA,GAG |
E236E |
NP_001034548.1 |
NM_001206480.2 |
3922 |
Silent Mutation |
GAA,GAG |
E716E |
NP_001193409.1 |
NM_001206482.1 |
3922 |
Silent Mutation |
GAA,GAG |
E716E |
NP_001193411.1 |
NM_014800.10 |
3922 |
Silent Mutation |
GAA,GAG |
E716E |
NP_055615.8 |
NM_130442.3 |
3922 |
Silent Mutation |
GAA,GAG |
E236E |
NP_569709.1 |
XM_005249919.2 |
3922 |
Silent Mutation |
GAA,GAG |
E716E |
XP_005249976.1 |
XM_006715805.1 |
3922 |
Silent Mutation |
GAA,GAG |
E716E |
XP_006715868.1 |
XM_011515654.1 |
3922 |
Silent Mutation |
GAA,GAG |
E716E |
XP_011513956.1 |
XM_017012838.1 |
3922 |
Silent Mutation |
GAA,GAG |
E716E |
XP_016868327.1 |
XM_017012839.1 |
3922 |
Silent Mutation |
GAA,GAG |
E716E |
XP_016868328.1 |
View Full Product Details