Product Details

SNP ID
rs138792415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73737078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGCGCCCGTCTACCTCTACCAG[A/G]TTAGTGAGCAGGTGCTGCCGCACGG
Phenotype
MIM: 612545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD11 PubMed Links

Gene Details

Gene
ABHD11
Gene Name
abhydrolase domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145364.2 833 Silent Mutation AAC,AAT N165N NP_001138836.1
NM_001301058.1 833 Intron NP_001287987.1
NM_001321382.1 833 Silent Mutation AAC,AAT N109N NP_001308311.1
NM_001321383.1 833 UTR 3 NP_001308312.1
NM_148912.3 833 Silent Mutation AAC,AAT N222N NP_683710.1
NM_148913.3 833 Silent Mutation AAC,AAT N215N NP_683711.1
XM_006716144.2 833 Silent Mutation AAC,AAT N109N XP_006716207.1
XM_011516590.2 833 Silent Mutation AAC,AAT N201N XP_011514892.1
XM_011516591.1 833 Silent Mutation AAC,AAT N109N XP_011514893.1
Gene
ABHD11-AS1
Gene Name
ABHD11 antisense RNA 1 (tail to tail)
There are no transcripts associated with this gene.

View Full Product Details